Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001162870.1 | NP_001156342.1 | 6 | 963 | |
NM_016962.2 | NP_058658.2 | 6 | 960 | ||
GenBank mRNA | AJ246002.1 | - | 6 | 585 | |
AK007793.1 | - | 6 | 740 | ||
AK044900.1 | - | 6 | 815 | ||
AK129282.1 | - | 6 | 937 | ||
BC046286.1 | - | 6 | 960 | 6441742 |