Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001093764.1 | NP_001087233.1 | 3 | 907 | |
NM_001093765.1 | NP_001087234.1 | 3 | 856 | ||
NM_001093766.1 | NP_001087235.1 | 2 | 794 | ||
NM_016969.2 | NP_058665.2 | 3 | 695 | ||
GenBank mRNA | AJ001616.1 | - | 3 | 716 | |
AK052258.1 | - | 3 | 814 | ||
AK053414.1 | - | 3 | 647 | ||
AK089538.1 | - | 3 | 678 | ||
AK149962.1 | - | 3 | 838 | ||
AK159160.1 | - | 838 | |||
AK180990.1 | - | 1 | 142 | ||
AK197378.1 | - | 1 | 154 | ||
BC056355.1 | - | 3 | 790 | 6811773 | |
BC119364.1 | - | 2 | 578 | 8734113 | |
BC119368.1 | - | 2 | 575 | 8734117 |