Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001166413.1 | NP_001159885.1 | 2 | 172 | |
NM_028027.3 | NP_082303.2 | 2 | 234 | ||
GenBank mRNA | AF487515.1 | - | 2 | 200 | |
AK010452.1 | - | 2 | 200 | ||
AK158684.1 | - | 2 | 172 | ||
BC041113.1 | - | 2 | 216 | ||
CF724487.1 | - | 2 | 245 |