Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026559.3 | NP_080835.1 | 2 | 246 | |
GenBank mRNA | AJ344103.1 | - | 2 | 235 | |
AK004219.1 | - | 2 | 254 | ||
AK007595.1 | - | 2 | 250 | ||
AK013103.1 | - | 2 | 189 | ||
AK076425.1 | - | 2 | 246 | ||
AK169882.1 | - | 2 | 189 | ||
BC030344.1 | - | 2 | 244 | 3673521 |