Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_173742.3 | NP_776103.1 | 1, 2 | 195 | |
GenBank mRNA | AK003548.1 | - | 1, 2 | 171 | |
AK009592.1 | - | 1, 2 | 190 | ||
AK018709.1 | - | 1, 2 | 144 | ||
AK171975.1 | - | 1, 2 | 144 | ||
BC021602.1 | - | 1, 2 | 123 | 5102170 | |
BC021603.1 | - | 1, 2 | 123 | 5102465 | |
BC024417.1 | - | 1, 2 | 153 | 5065541 | |
CJ181610.1 | - | 1, 2 | 195 |