Gene Transcripts
- Gene Symbol
- Cdkl2
- Entrez Gene ID
- 53886
- Gene Name
- cyclin-dependent kinase-like 2 (CDC2-related kinase)
- Gene Aliases
- 5330436L21Rik, AI505225, KKIAMRE, Kkm
- Chromosome Location
- Chr. 5: 92006074 - 92043906 on Build GRCm38
- UniGene ID
- Mm.44963
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001276315.1 |
NP_001263244.1
|
10, 11 |
1958 |
|
NM_016912.2 |
NP_058608.1
|
10, 11 |
1692 |
|
NM_177270.4 |
NP_796244.2
|
10, 11 |
1692 |
|
GenBank mRNA |
AB029065.1 |
- |
10, 11 |
1668 |
|
AB029066.1 |
- |
10, 11 |
1656 |
|
AK030598.1 |
- |
10, 11 |
1947 |
|
AK139820.1 |
- |
10, 11 |
1901 |
|
AK144574.1 |
- |
10, 11 |
1693 |
|
BC117894.1 |
- |
10, 11 |
1601 |
40096852 |
BC117895.1 |
- |
10, 11 |
1601 |
40096853 |
View Full Product Details