Gene Transcripts
- Gene Symbol
- Snx3
- Entrez Gene ID
- 54198
- Gene Name
- sorting nexin 3
- Gene Aliases
- SDP3
- Chromosome Location
- Chr. 10: 42502054 - 42535369 on Build GRCm38
- UniGene ID
- Mm.277870
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_017472.4 |
NP_059500.2
|
1, 2 |
434 |
|
GenBank mRNA |
AF062482.1 |
- |
1, 2 |
232 |
|
AK007798.1 |
- |
1, 2 |
245 |
|
AK013183.1 |
- |
1, 2 |
459 |
|
AK013261.1 |
- |
1, 2 |
67 |
|
AK018790.1 |
- |
1, 2 |
67 |
|
AK145350.1 |
- |
1, 2 |
237 |
|
AK154728.1 |
- |
1, 2 |
237 |
|
AK189751.1 |
- |
1, 2 |
25 |
|
BC116885.1 |
- |
1, 2 |
278 |
40126204 |
BC116889.1 |
- |
1, 2 |
278 |
40126208 |
View Full Product Details