Gene Transcripts

Gene Symbol
Chst2
Entrez Gene ID
54371
Gene Name
carbohydrate sulfotransferase 2
Gene Aliases
AI428561, AW121776, C130041E03Rik, Chts2, GST-2, GlcNAc6ST, Gn6st
Chromosome Location
Chr. 9: 95400926 - 95407270 on Build GRCm38
UniGene ID
Mm.212446
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018763.2 NP_061233.2 2 1764
GenBank mRNA AB011451.1 - 2 1216
AB011452.1 - 2 1216
BC051963.1 - 2 1151 5707543

View Full Product Details