Gene Transcripts

Gene Symbol
Sfn
Entrez Gene ID
55948
Gene Name
stratifin
Gene Aliases
Er, Mme1, Ywhas
Chromosome Location
Chr. 4: 133600556 - 133602168 on Build GRCm38
UniGene ID
Mm.44482
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018754.2 NP_061224.2 1 924
GenBank mRNA AF058798.1 - 1 688
AF152893.1 - 1 525
AK146490.1 - 1 598
AK169358.1 - 1 589

View Full Product Details