Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_000422.2 | NP_000413.1 | 1 | 536 | |
GenBank mRNA | AK095342.1 | - | 1 | 465 | |
AK304373.1 | - | 1 | 483 | ||
BC000159.2 | - | 1 | 465 | 2986100 | |
BC011901.2 | - | 1 | 463 | 4111703 | |
BC056421.1 | - | 1 | 465 | 5208304 | |
BC072018.1 | - | 2 | 403 | 3531173 | |
BC072019.1 | - | 1 | 461 | 5442690 | |
BP226061.1 | - | 1 | 536 | ||
BX647923.1 | - | 1 | 202 | ||
X62571.1 | - | 1 | 463 |