Gene Transcripts

Gene Symbol
Cpxm2
Entrez Gene ID
55987
Gene Name
carboxypeptidase X 2 (M14 family)
Gene Aliases
4632435C11Rik, CPX-2, Cpx2
Chromosome Location
Chr. 7: 132032687 - 132154768 on Build GRCm38
UniGene ID
Mm.471654
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_018867.5 NP_061355.3 11, 12 1930
GenBank mRNA AF017639.1 - 11, 12 1931
AK019509.1 - 11, 12 1929
BC021444.1 - 11, 12 1852 5038528

View Full Product Details