Gene Transcripts

Gene Symbol
Fscn3
Entrez Gene ID
56223
Gene Name
fascin homolog 3, actin-bundling protein, testicular (Strongylocentrotus purpuratus)
Gene Aliases
Chromosome Location
Chr. 6: 28427870 - 28439398 on Build GRCm38
UniGene ID
Mm.31708
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019569.2 NP_062515.2 2 798
GenBank mRNA AF176024.2 - 2 910
BC050759.1 - 2 798 6770861

View Full Product Details