Gene Transcripts

Gene Symbol
Ppp4c
Entrez Gene ID
56420
Gene Name
protein phosphatase 4, catalytic subunit
Gene Aliases
1110002D08Rik, AU016079, Ppx
Chromosome Location
Chr. 7: 126785866 - 126792471 on Build GRCm38
UniGene ID
Mm.41998
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019674.3 NP_062648.1 4 291
GenBank mRNA AF088911.1 - 4 316
AK027923.1 - 2 70
AK144244.1 - 3 218
BC001993.1 - 4 291 3491568

View Full Product Details