Gene Transcripts

Gene Symbol
Spata5
Entrez Gene ID
57815
Gene Name
spermatogenesis associated 5
Gene Aliases
2510048F20Rik, C78064, Spaf
Chromosome Location
Chr. 3: 37420280 - 37579096 on Build GRCm38
UniGene ID
Mm.172679
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001163511.1 NP_001156983.1 2 236
NM_021343.2 NP_067318.2 2 233
GenBank mRNA AF049099.1 - 2 215
AK011111.2 - 2 200
AK144998.1 - 2 563
BC145302.1 - 2 204 9053856
BC145309.1 - 2 204
BY106106.1 - 2 237

View Full Product Details