Gene Transcripts

Gene Symbol
Keg1
Entrez Gene ID
64697
Gene Name
kidney expressed gene 1
Gene Aliases
0610008P16Rik, AI316519, GS4059
Chromosome Location
Chr. 19: 12695790 - 12719902 on Build GRCm38
UniGene ID
Mm.218561
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_029550.4 NP_083826.1 6 789
GenBank mRNA AB028071.1 - 6 766
AK002360.1 - 6 787
AK165515.1 - 6 787
BC010803.1 - 6 779 4238654

View Full Product Details