Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_022992.2 | NP_075368.1 | 3 | 519 | |
GenBank mRNA | AF265214.1 | - | 3 | 490 | |
AK005259.1 | - | 3 | 516 | ||
AK008519.1 | - | 3 | 499 | ||
AK076519.1 | - | 3 | 516 | ||
AK077826.1 | - | 3 | 525 | ||
AK088205.1 | - | 3 | 514 | ||
BC003897.1 | - | 3 | 519 | 3600236 | |
D87211.1 | - | 3 | 562 |