Gene Transcripts

Gene Symbol
Lsm7
Entrez Gene ID
66094
Gene Name
LSM7 homolog, U6 small nuclear RNA associated (S. cerevisiae)
Gene Aliases
0910001B06Rik, 1110033F18Rik
Chromosome Location
Chr. 10: 80852823 - 80855249 on Build GRCm38
UniGene ID
Mm.379101
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025349.2 NP_079625.1 2 82
GenBank mRNA AK003062.1 - 2 70
AK007462.1 - 2 79
AK012446.1 - 2 79
AK013355.1 - 2 83
BC027511.1 - 2 72 1382565
BC081444.1 - 1 65 5711629

View Full Product Details