Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025570.2 | NP_079846.1 | 2, 3 | 244 | |
GenBank mRNA | AB049645.1 | - | 2, 3 | 217 | |
AK004053.1 | - | 2, 3 | 229 | ||
AK011341.1 | - | 2, 3 | 245 | ||
AK015199.1 | - | 228 | |||
AK168665.1 | - | 2, 3 | 218 | ||
BC016591.1 | - | 2, 3 | 207 | 3158162 | |
BC039985.1 | - | 2, 3 | 214 | 1244957 |