Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025610.3 | NP_079886.2 | 3, 4 | 405 | |
GenBank mRNA | AK006147.1 | - | 3, 4 | 441 | |
AK010381.1 | - | 3, 4 | 447 | ||
AK030209.1 | - | 3, 4 | 406 | ||
BC016106.1 | - | 1, 2 | 157 | 4921265 |