Gene Transcripts

Gene Symbol
Krtap3-2
Entrez Gene ID
66708
Gene Name
keratin associated protein 3-2
Gene Aliases
2310020D06Rik, 4733401L16Rik, KAP3.2, KRTAP3.2
Chromosome Location
Chr. 11: 99555823 - 99556853 on Build GRCm38
UniGene ID
Mm.46389
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025720.3 NP_079996.2 1 561
GenBank mRNA AK003615.1 - 1 563
AK014641.1 - 1 562
BY703877.1 - 1 563

View Full Product Details