Gene Transcripts

Gene Symbol
Kcng4
Entrez Gene ID
66733
Gene Name
potassium voltage-gated channel, subfamily G, member 4
Gene Aliases
4921535I01Rik, AW049024, KV6.3, KV6.4
Chromosome Location
Chr. 8: 119623854 - 119636353 on Build GRCm38
UniGene ID
Mm.358699
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025734.2 NP_080010.2 3 1652
GenBank mRNA AF450109.1 - 3 1318
AK015001.1 - 1 119
BC043936.1 - 3 1652 4504272

View Full Product Details