Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_025770.3 | NP_080046.3 | 2 | 101 | |
GenBank mRNA | AB079383.1 | - | 1 | 57 | |
AB091691.1 | - | 1 | 66 | ||
AK009076.1 | - | 2 | 85 | ||
AK017355.1 | - | 2 | 83 | ||
AK077333.1 | - | 2 | 74 | ||
BB843097.1 | - | 2 | 101 | ||
BC023455.1 | - | 1 | 65 | 5249268 |