Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_023289.2 | NP_075778.2 | 2 | 298 | |
GenBank mRNA | AF221099.1 | - | 2 | 341 | |
AK004142.1 | - | 2 | 299 | ||
AK005432.1 | - | 2 | 299 | ||
AK005463.1 | - | 2 | 299 | ||
AK145847.1 | - | 2 | 299 | ||
BC099452.1 | - | 2 | 286 | 30787372 |