Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001145780.1 | NP_001139252.1 | 4 | 457 | |
NM_025917.4 | NP_080193.1 | 3 | 244 | ||
GenBank mRNA | AF353245.1 | - | 3 | 237 | |
AK003257.1 | - | 3 | 237 | ||
AK003385.1 | - | 2 | 108 | ||
AK008573.1 | - | 3 | 237 | ||
AK008661.1 | - | 2 | 142 | ||
AK012490.1 | - | 3 | 238 | ||
AK017484.1 | - | 1 | 57 | ||
AK160726.1 | - | 3 | 237 | ||
AK165639.1 | - | 3 | 386 | ||
AY842453.1 | - | 3 | 223 | ||
BC025086.1 | - | 2 | 320 | 3995001 | |
BC038519.1 | - | 2 | 304 | 1428854 | |
BC075695.1 | - | 4 | 363 | 30623689 |