Gene Transcripts

Gene Symbol
Vma21
Entrez Gene ID
67048
Gene Name
VMA21 vacuolar H+-ATPase homolog (S. cerevisiae)
Gene Aliases
2610030H06Rik, AI840175
Chromosome Location
Chr. X: 71812752 - 71824706 on Build GRCm38
UniGene ID
Mm.371816
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001081356.3 NP_001074825.1 2 267
NM_001290780.1 NP_001277709.1 2 394
NM_001290781.1 NP_001277710.1 3 568
GenBank mRNA AA015086.1 - 3 336
AK008848.1 - 2 225
AK011632.1 - 2 233
AK045401.1 - 2 225
AK049296.1 - 2 205
AK049488.1 - 2 238
AK138582.1 - 235
AK164163.1 - 2 222
BC028317.1 - 2 205
BC094639.1 - 2 221 1361961
CJ062712.1 - 2 247
CJ071023.1 - 2 267

View Full Product Details