Gene Transcripts
- Gene Symbol
- Rnpc3
- Entrez Gene ID
- 67225
- Gene Name
- RNA-binding region (RNP1, RRM) containing 3
- Gene Aliases
- 2810441O16Rik, AI447568, C030014B17Rik
- Chromosome Location
- Chr. 3: 113605067 - 113630149 on Build GRCm38
- UniGene ID
- Mm.316928
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001038696.1 |
NP_001033785.1
|
12, 13 |
1448 |
|
NM_001287015.1 |
NP_001273944.1
|
12, 13 |
1448 |
|
NM_026043.3 |
NP_080319.2
|
12, 13 |
1448 |
|
GenBank mRNA |
AK134241.1 |
- |
12, 13 |
1413 |
|
AK161063.1 |
- |
12, 13 |
1431 |
|
AK161974.1 |
- |
12, 13 |
1400 |
|
AK163729.1 |
- |
12, 13 |
1400 |
|
AK173272.1 |
- |
12, 13 |
1199 |
|
BC016603.1 |
- |
9, 10 |
964 |
3587176 |
BC043695.1 |
- |
12, 13 |
1417 |
5250774 |
View Full Product Details