Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001162523.1 | NP_001155995.1 | 3 | 849 | |
NM_001162524.1 | NP_001155996.1 | 3 | 849 | ||
NM_026087.3 | NP_080363.2 | 3 | 849 | ||
GenBank mRNA | AK005400.1 | - | 3 | 851 | |
AK005565.1 | - | 3 | 800 | ||
AK131656.1 | - | 3 | 839 | ||
AK145926.1 | - | 3 | 800 | ||
BC099443.1 | - | 3 | 787 | 30787251 |