Gene Transcripts

Gene Symbol
Narf
Entrez Gene ID
67608
Gene Name
nuclear prelamin A recognition factor
Gene Aliases
4430402O11Rik
Chromosome Location
Chr. 11: 121237236 - 121255856 on Build GRCm38
UniGene ID
Mm.291832
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026272.3 NP_080548.3 5 522
GenBank mRNA AK013432.1 - 5 481
AK076232.1 - 5 505
AK081305.1 - 5 506
BC016090.1 - 5 461 4503713

View Full Product Details