Gene Transcripts

Gene Symbol
Ttc39d
Entrez Gene ID
67737
Gene Name
tetratricopeptide repeat domain 39D
Gene Aliases
4930560E09Rik
Chromosome Location
Chr. 17: 80215914 - 80217936 on Build GRCm38
UniGene ID
Mm.158172
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026351.2 NP_080627.2 1 400
GenBank mRNA BC118946.1 - 1 400
BC118947.1 - 1 400 40092368

View Full Product Details