Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026385.4 | NP_080661.1 | 2, 3 | 379 | |
GenBank mRNA | AK002477.1 | - | 2, 3 | 380 | |
AK018198.1 | - | 2, 3 | 368 | ||
AK032540.1 | - | 2, 3 | 368 | ||
BC024534.1 | - | 2, 3 | 370 | 4987882 |