Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_026467.3 | NP_080743.1 | 2, 3 | 180 | |
GenBank mRNA | AK007689.1 | - | 2, 3 | 181 | |
AK011262.1 | - | 2, 3 | 199 | ||
AK151083.1 | - | 2, 3 | 199 | ||
BC058115.1 | - | 2, 3 | 181 | 6407738 |