Gene Transcripts

Gene Symbol
Trabd
Entrez Gene ID
67976
Gene Name
TraB domain containing
Gene Aliases
5730502D15Rik, AL023039
Chromosome Location
Chr. 15: 89076050 - 89087078 on Build GRCm38
UniGene ID
Mm.284710
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026485.2 NP_080761.2 8 871
GenBank mRNA AK005300.1 - 8 841
AK017745.1 - 8 865
AK131116.1 - 1 2835
AK146027.1 - 8 887
BC005574.1 - 8 897 3967961

View Full Product Details