Gene Transcripts
- Gene Symbol
- Smarcc2
- Entrez Gene ID
- 68094
- Gene Name
- SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily c, member 2
- Gene Aliases
- 5930405J04Rik
- Chromosome Location
- Chr. 10: 128459236 - 128490583 on Build GRCm38
- UniGene ID
- Mm.417338
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001114096.1 |
NP_001107568.1
|
10, 11 |
1058 |
|
NM_001114097.1 |
NP_001107569.1
|
10, 11 |
1058 |
|
NM_198160.2 |
NP_937803.1
|
10, 11 |
1058 |
|
GenBank mRNA |
AK089461.1 |
- |
10, 11 |
994 |
|
AK146971.1 |
- |
10, 11 |
987 |
|
AK170530.1 |
- |
10, 11 |
993 |
|
AK215230.1 |
- |
2, 3 |
166 |
|
BC040363.1 |
- |
10, 11 |
973 |
|
BC053452.1 |
- |
1, 2 |
119 |
|
BC058720.1 |
- |
10, 11 |
1000 |
5705438 |
View Full Product Details