Gene Transcripts

Gene Symbol
Nrn1
Entrez Gene ID
68404
Gene Name
neuritin 1
Gene Aliases
0710008J23Rik, Cpg15, Nrn
Chromosome Location
Chr. 13: 36725622 - 36734477 on Build GRCm38
UniGene ID
Mm.232930
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_153529.2 NP_705757.1 2, 3 380
GenBank mRNA AK003046.1 - 2, 3 207
AK161859.1 - 2, 3 348
BC035531.1 - 2, 3 368 5367281

View Full Product Details