Gene Transcripts

Gene Symbol
Tomm5
Entrez Gene ID
68512
Gene Name
translocase of outer mitochondrial membrane 5 homolog (yeast)
Gene Aliases
1110019J04Rik, Tom5
Chromosome Location
Chr. 4: 45105209 - 45108113 on Build GRCm38
UniGene ID
Mm.282081
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099675.1 NP_001093145.1 2 587
NM_001134646.1 NP_001128118.1 3 637
GenBank mRNA AK003822.1 - 2 583
CA466100.1 - 3 629

View Full Product Details