Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_024229.2 | NP_077191.2 | 12 | 1026 | |
GenBank mRNA | AK140458.1 | - | 12 | 1066 | |
AK144544.1 | - | 12 | 1075 | ||
AK166319.1 | - | 11 | 1001 | ||
BC003473.1 | - | 12 | 1032 | 2650077 | |
BC008276.1 | - | 12 | 1026 | 3707732 |