Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_178874.2 | NP_849205.1 | 3 | 1095 | |
GenBank mRNA | AK145948.1 | - | 2 | 456 | |
AK157378.1 | - | 3 | 1208 | ||
AK158436.1 | - | 2 | 457 | ||
AK165309.1 | - | 2 | 456 | ||
AK171635.1 | - | 2 | 456 | ||
AK172313.1 | - | 2 | 456 | ||
AK172356.1 | - | 2 | 456 | ||
AK172382.1 | - | 2 | 456 | ||
AK172541.1 | - | 2 | 456 | ||
AK172720.1 | - | 2 | 456 | ||
AK172957.1 | - | 3 | 950 | ||
BC052035.1 | - | 3 | 1095 | 5695005 |