Gene Transcripts

Gene Symbol
Ptcd2
Entrez Gene ID
68927
Gene Name
pentatricopeptide repeat domain 2
Gene Aliases
1190005P08Rik
Chromosome Location
Chr. 13: 99319649 - 99344678 on Build GRCm38
UniGene ID
Mm.276502
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026873.2 NP_081149.1 6 586
GenBank mRNA AK004502.1 - 6 596
AK144396.1 - 6 598
BC016563.1 - 6 588
BC025110.1 - 6 586 3987809

View Full Product Details