Gene Transcripts

Gene Symbol
Srrm4
Entrez Gene ID
68955
Gene Name
serine/arginine repetitive matrix 4
Gene Aliases
1500001A10Rik, B230202K19Rik, bv, mKIAA1853, nSR100
Chromosome Location
Chr. 5: 116439272 - 116592562 on Build GRCm38
UniGene ID
Mm.146779
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_026886.3 NP_081162.1 6 829
GenBank mRNA AK053836.1 - 6 561
AK165719.1 - 6 547
BC056392.1 - 6 819 6855144
JN860221.1 - 6 476

View Full Product Details