Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_027000.4 | NP_081276.2 | 2 | 116 | |
GenBank mRNA | AF325354.1 | - | 2 | 100 | |
AF348208.1 | - | 2 | 123 | ||
AK011580.1 | - | 2 | 127 | ||
AK141852.1 | - | 2 | 126 | ||
AK164330.1 | - | 2 | 117 | ||
AK166711.1 | - | 2 | 122 | ||
BC005514.1 | - | 2 | 97 | ||
BC025431.1 | - | 2 | 121 | 5007174 |