Gene Transcripts
- Gene Symbol
- Ttl
- Entrez Gene ID
- 69737
- Gene Name
- tubulin tyrosine ligase
- Gene Aliases
- 2410003M22Rik, 2700049H19Rik, AI848570
- Chromosome Location
- Chr. 2: 129065947 - 129096283 on Build GRCm38
- UniGene ID
- Mm.33909
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC018513.1 |
- |
5 |
841 |
|
BC024414.1 |
- |
5 |
810 |
4973810 |
View Full Product Details