Gene Transcripts

Gene Symbol
Zdhhc25
Entrez Gene ID
70073
Gene Name
zinc finger, DHHC domain containing 25
Gene Aliases
1700030J15Rik
Chromosome Location
Chr. 15: 88600302 - 88601669 on Build GRCm38
UniGene ID
Mm.45306
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027306.3 NP_081582.1 1 598
GenBank mRNA AK006552.1 - 1 586
AK131706.1 - 1 572
BC049767.1 - 1 598 6774409

View Full Product Details