Gene Transcripts
- Gene Symbol
- MN1
- Entrez Gene ID
- 4330
- Gene Name
- MN1 proto-oncogene, transcriptional regulator
- Gene Aliases
- MGCR, MGCR1, MGCR1-PEN, dJ353E16.2
- Chromosome Location
- Chr. 22: 27748277 - 27801498 on Build GRCh38
- UniGene ID
- Hs.268515
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_002430.2 |
NP_002421.3
|
1 |
1686 |
|
GenBank mRNA |
X82209.2 |
- |
1 |
1687 |
|
View Full Product Details