Gene Transcripts

Gene Symbol
Speer4f
Entrez Gene ID
70935
Gene Name
spermatogenesis associated glutamate (E)-rich protein 4f
Gene Aliases
4922502J04Rik, SPEER-4F
Chromosome Location
Chr. 5: 17476098 - 17480936 on Build GRCm38
UniGene ID
Mm.332225
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027609.2 NP_081885.1 4 590
GenBank mRNA AK015022.1 - 4 593
BC116901.1 - 4 616 40126220
BC116903.1 - 4 616 40126222

View Full Product Details