Gene Transcripts

Gene Symbol
Fam83c
Entrez Gene ID
71405
Gene Name
family with sequence similarity 83, member C
Gene Aliases
5530400B04Rik
Chromosome Location
Chr. 2: 155829183 - 155834854 on Build GRCm38
UniGene ID
Mm.260918
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027788.2 NP_082064.2 3 805
GenBank mRNA BC113190.1 - 3 804
BC113797.1 - 3 804 40062621

View Full Product Details