Gene Transcripts

Gene Symbol
Alpk1
Entrez Gene ID
71481
Gene Name
alpha-kinase 1
Gene Aliases
8430410J10Rik, LAK
Chromosome Location
Chr. 3: 127670308 - 127780553 on Build GRCm38
UniGene ID
Mm.158704
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_027808.1 NP_082084.1 3 715
GenBank mRNA AK018401.1 - 3 685
AK041315.1 - 3 702
AK173191.1 - 3 715

View Full Product Details