Gene Transcripts

Gene Symbol
Sfpq
Entrez Gene ID
71514
Gene Name
splicing factor proline/glutamine rich (polypyrimidine tract binding protein associated)
Gene Aliases
1110004P21Rik, 2810416M14Rik, 5730453G22Rik, 9030402K04Rik, AU021830, D4Ertd314e, Gm12940, OTTMUSG00000009329, PSF, REP1
Chromosome Location
Chr. 4: 127020560 - 127037016 on Build GRCm38
UniGene ID
Mm.257276
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023603.3 NP_076092.1 8, 9 1963
GenBank mRNA AK018483.1 - 3, 4 256
AK191209.1 - 1, 2 45
AK192184.1 - 1, 2 45
AK198254.1 - 1, 2 45
AK211103.1 - 1, 2 45
AK217233.1 - 2, 3 125
AK219185.1 - 1, 2 45
AY034062.1 - 8, 9 1878
BC037486.1 - 6, 7 676
BC049227.1 - 5, 6 534
BC055468.1 - 4, 5 321
BC080848.1 - 8, 9 1929
BC089305.1 - 8, 9 1929 30360144
CA319647.1 - 6, 7 594
CJ304628.1 - 2, 3 153

View Full Product Details