Gene Transcripts

Gene Symbol
Sfpq
Entrez Gene ID
71514
Gene Name
splicing factor proline/glutamine rich (polypyrimidine tract binding protein associated)
Gene Aliases
1110004P21Rik, 2810416M14Rik, 5730453G22Rik, 9030402K04Rik, AU021830, D4Ertd314e, Gm12940, OTTMUSG00000009329, PSF, REP1
Chromosome Location
Chr. 4: 127020560 - 127037016 on Build GRCm38
UniGene ID
Mm.257276
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_023603.3 NP_076092.1 7, 8 1900
GenBank mRNA AK018483.1 - 2, 3 193
AK217233.1 - 1, 2 62
AY034062.1 - 7, 8 1815
BC037486.1 - 5, 6 613
BC049227.1 - 4, 5 471
BC055468.1 - 3, 4 258
BC080848.1 - 7, 8 1866
BC089305.1 - 7, 8 1866 30360144
CA319647.1 - 5, 6 531
CJ304628.1 - 1, 2 90

View Full Product Details