Gene Transcripts

Gene Symbol
MSI1
Entrez Gene ID
4440
Gene Name
musashi RNA binding protein 1
Gene Aliases
Chromosome Location
Chr. 12: 120339655 - 120369180 on Build GRCh38
UniGene ID
Hs.158311
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002442.3 NP_002433.1 6 475
XM_006719403.3 XP_006719466.1 6 469
XM_011538361.2 XP_011536663.1 6 469
XM_011538362.2 XP_011536664.1 6 469
XM_011538363.2 XP_011536665.1 6 469
XM_011538364.2 XP_011536666.1 2 1019
XM_011538365.2 XP_011536667.1 3 138
XM_011538366.2 XP_011536668.1 3 1146
GenBank mRNA AB012851.1 - 6 450
AB072590.1 - 3 197
AB072591.1 - 3 197
AB072592.1 - 3 197
AB073212.1 - 3 190
BX393884.2 - 6 475

View Full Product Details