Gene Transcripts
- Gene Symbol
- Trub1
- Entrez Gene ID
- 72133
- Gene Name
- TruB pseudouridine (psi) synthase homolog 1 (E. coli)
- Gene Aliases
- 2610009I02Rik, 9030425C13Rik, AI448235, BB118943, C76870
- Chromosome Location
- Chr. 19: 57452891 - 57491005 on Build GRCm38
- UniGene ID
- Mm.258404
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_028115.3 |
NP_082391.1
|
4, 5 |
526 |
|
NM_029839.3 |
NP_084115.2
|
4, 5 |
526 |
|
GenBank mRNA |
AK011362.1 |
- |
4, 5 |
521 |
|
AK018525.1 |
- |
4, 5 |
499 |
|
AK031674.1 |
- |
4, 5 |
504 |
|
AK164095.1 |
- |
4, 5 |
497 |
|
AK169221.1 |
- |
4, 5 |
492 |
|
BC039287.1 |
- |
4, 5 |
471 |
|
BC054088.1 |
- |
4, 5 |
340 |
|
BC089592.1 |
- |
4, 5 |
493 |
6757213 |
View Full Product Details